The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138693.3 | NP_619638.2 | 1 - 1 | 853 | 76 |
GenBank mRNA | AF490374.1 | - | 1 - 1 | 853 | 76 |
DQ534757.1 | - | 1 - 1 | 927 | 76 |