Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TAS1R1
Entrez Gene ID
80835
Gene Name
taste 1 receptor member 1
Gene Aliases
GM148, GPR70, T1R1, TR1
Chromosome Location
Chr.1: 6555278 - 6579757 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138697.3 NP_619642.2 2 - 3 593 81
XM_011542203.2 XP_011540505.1 2 - 3 959 81
XM_011542206.2 XP_011540508.1 1 - 2 1145 81
XM_017002402.1 XP_016857891.1 1 - 2 1145 81
GenBank mRNA AF387618.1 - 1 - 2 306 81
AK315805.1 - 2 - 3 562 81
BC136515.1 - 2 - 3 593 81
BC136516.1 - 2 - 3 592 81

View Full Product Details