The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006833.4 | NP_006824.2 | 5 - 6 | 516 | 99 |
GenBank mRNA | AK312024.1 | - | 5 - 6 | 507 | 99 |
BC002520.2 | - | 5 - 6 | 490 | 99 | |
CB990467.1 | - | 5 - 6 | 540 | 99 | |
U70735.1 | - | 5 - 6 | 438 | 99 |