Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BCOR
Entrez Gene ID
54880
Gene Name
BCL6 corepressor
Gene Aliases
ANOP2, MAA2, MCOPS2
Chromosome Location
Chr.X: 40051246 - 40177390 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123383.1 NP_001116855.1 5 - 6 3412 91
NM_001123385.1 NP_001116857.1 5 - 6 3341 91
NM_017745.5 NP_060215.4 5 - 6 3341 91
XM_005272616.1 XP_005272673.1 5 - 6 3473 91
XM_005272618.3 XP_005272675.1 5 - 6 3139 91
XM_006724536.3 XP_006724599.1 5 - 6 4575 91
XM_011543929.2 XP_011542231.1 5 - 6 3249 91
XM_011543930.1 XP_011542232.1 5 - 6 3262 91
XM_011543931.2 XP_011542233.1 5 - 6 3372 91
XM_017029615.1 XP_016885104.1 5 - 6 4575 91
XM_017029616.1 XP_016885105.1 5 - 6 3799 91
GenBank mRNA AF317391.1 - 5 - 6 3300 91
AK056538.1 - 5 - 6 3166 91
AY316592.1 - 5 - 6 3300 91

View Full Product Details