The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030957.3 | NP_112219.3 | 5 - 6 | 899 | 86 |
XM_006722917.3 | XP_006722980.1 | 4 - 5 | 740 | 86 | |
XM_017027338.1 | XP_016882827.1 | 4 - 5 | 755 | 86 | |
GenBank mRNA | AB209515.1 | - | 4 - 5 | 730 | 86 |
AF163762.1 | - | 2 - 3 | 515 | 86 | |
BX461830.2 | - | 4 - 5 | 678 | 86 |