The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001184714.1 | NP_001171643.1 | 2 - 3 | 452 | 99 |
NM_001184715.1 | NP_001171644.1 | 2 - 3 | 305 | 99 | |
NM_052931.4 | NP_443163.1 | 2 - 3 | 452 | 99 | |
XM_017000215.1 | XP_016855704.1 | 2 - 3 | 458 | 99 | |
XM_017000216.1 | XP_016855705.1 | 2 - 3 | 278 | 99 | |
GenBank mRNA | AJ277141.1 | - | 2 - 3 | 394 | 99 |
AJ306388.1 | - | 2 - 3 | 391 | 99 | |
AK125624.1 | - | 2 - 3 | 452 | 99 | |
AK303990.1 | - | 2 - 3 | 298 | 99 | |
AK310085.1 | - | 2 - 3 | 280 | 99 | |
AK313549.1 | - | 2 - 3 | 404 | 99 | |
AL832854.1 | - | 2 - 3 | 433 | 99 | |
AY358159.1 | - | 2 - 3 | 443 | 99 | |
BC113893.1 | - | 2 - 3 | 409 | 99 | |
BC114495.1 | - | 2 - 3 | 409 | 99 | |
DB197632.1 | - | 2 - 3 | 452 | 99 |