Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NRXN1
Entrez Gene ID
9378
Gene Name
neurexin 1
Gene Aliases
Hs.22998, PTHSL2, SCZD17
Chromosome Location
Chr.2: 49918503 - 51032536 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138735.2 NP_620072.1 1 - 2 1069 74
XM_017005335.1 XP_016860824.1 1 - 2 6059 74
XM_017005336.1 XP_016860825.1 1 - 2 6060 74
XM_017005337.1 XP_016860826.1 1 - 2 6059 74
GenBank mRNA AF064842.1 - 1 - 2 1068 74
BC125179.1 - 2 - 3 125 74
BC125180.1 - 2 - 3 488 74

View Full Product Details