Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NFATC2IP
Entrez Gene ID
84901
Gene Name
nuclear factor of activated T-cells 2 interacting protein
Gene Aliases
ESC2, NIP45, RAD60
Chromosome Location
Chr.16: 28950988 - 28967097 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032815.3 NP_116204.3 4 - 5 674 68
GenBank mRNA AF458593.1 - 4 - 5 684 68
AK027545.1 - 1 - 2 89 68
AK074761.1 - 4 - 5 664 68
BC018311.1 - 2 - 3 178 68
BC021551.1 - 1 - 2 57 68
BC080628.1 - 4 - 5 341 68
BC101741.1 - 4 - 5 734 68
BC112182.1 - 4 - 5 734 68

View Full Product Details