The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318976.1 | NP_001305905.1 | 6 - 7 | 626 | 91 |
NM_001318977.1 | NP_001305906.1 | 5 - 6 | 496 | 91 | |
NM_005680.2 | NP_005671.2 | 5 - 6 | 496 | 91 | |
XM_011510410.2 | XP_011508712.1 | 5 - 6 | 437 | 91 | |
XM_011510411.1 | XP_011508713.1 | 6 - 7 | 545 | 91 | |
GenBank mRNA | AK026521.1 | - | 5 - 6 | 413 | 91 |
AK295402.1 | - | 6 - 7 | 545 | 91 | |
AK308070.1 | - | 5 - 6 | 445 | 91 | |
AK309751.1 | - | 5 - 6 | 479 | 91 | |
AK315098.1 | - | 5 - 6 | 428 | 91 | |
BC018137.2 | - | 5 - 6 | 495 | 91 | |
L39061.1 | - | 5 - 6 | 498 | 91 |