Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 1771188-1771698 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLN8
Entrez Gene ID
2055
Gene Name
ceroid-lipofuscinosis, neuronal 8
Gene Aliases
C8orf61, EPMR
UniGene
Hs.127675
Gene Chromosome Location
Chr.8: 1755778-1786570 on Build GRCh38
Transcripts
NM_018941.3 XM_005266021.3 XM_005266022.1 XM_005266023.1 XM_011534745.1 XM_011534746.2 XM_011534747.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3898495 c.333G>A p.T111T Chr.8: 1771387-1771387 on Build GRCh38
6630572 c.288G>A p.A96A Chr.8: 1771342-1771342 on Build GRCh38
6630570 c.471C>G p.H157Q Chr.8: 1771525-1771525 on Build GRCh38
7195887 c.304_337del34 p.W102fs*14 Chr.8: 1771358-1771391 on Build GRCh38
6630578 c.331A>G p.T111A Chr.8: 1771385-1771385 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs796069556 Chr.8: 1771228-1771228 on Build GRCh38
rs772106347 Chr.8: 1771549-1771549 on Build GRCh38
rs751546464 Chr.8: 1771347-1771347 on Build GRCh38
rs116605307 Chr.8: 1771344-1771344 on Build GRCh38
rs756267448 Chr.8: 1771598-1771598 on Build GRCh38
rs750103399 Chr.8: 1771308-1771308 on Build GRCh38
rs794727299 Chr.8: 1771581-1771581 on Build GRCh38
rs919541697 Chr.8: 1771320-1771320 on Build GRCh38
rs768531216 Chr.8: 1771540-1771540 on Build GRCh38
rs140934218 Chr.8: 1771407-1771407 on Build GRCh38
rs372268977 Chr.8: 1771215-1771215 on Build GRCh38
rs148988002 Chr.8: 1771354-1771354 on Build GRCh38
rs776074645 Chr.8: 1771643-1771643 on Build GRCh38
rs770147696 Chr.8: 1771630-1771630 on Build GRCh38
rs746025062 Chr.8: 1771523-1771523 on Build GRCh38
rs554042394 Chr.8: 1771452-1771452 on Build GRCh38
rs778188757 Chr.8: 1771386-1771386 on Build GRCh38
rs746051539 Chr.8: 1771418-1771418 on Build GRCh38
rs762984374 Chr.8: 1771265-1771265 on Build GRCh38
rs184988046 Chr.8: 1771372-1771372 on Build GRCh38
rs772292767 Chr.8: 1771644-1771644 on Build GRCh38
rs779366141 Chr.8: 1771316-1771316 on Build GRCh38
rs376910635 Chr.8: 1771567-1771567 on Build GRCh38
rs775400768 Chr.8: 1771341-1771341 on Build GRCh38
rs143701028 Chr.8: 1771377-1771377 on Build GRCh38
rs759153401 Chr.8: 1771564-1771564 on Build GRCh38
rs386834127 Chr.8: 1771469-1771469 on Build GRCh38
rs759830733 Chr.8: 1771337-1771337 on Build GRCh38
rs779967066 Chr.8: 1771604-1771604 on Build GRCh38
rs1057516582 Chr.8: 1771317-1771317 on Build GRCh38
rs372313992 Chr.8: 1771333-1771333 on Build GRCh38
rs146424314 Chr.8: 1771322-1771322 on Build GRCh38
rs370471193 Chr.8: 1771614-1771614 on Build GRCh38
rs144015378 Chr.8: 1771300-1771300 on Build GRCh38
rs777822558 Chr.8: 1771636-1771636 on Build GRCh38
rs974289528 Chr.8: 1771634-1771634 on Build GRCh38
rs147224140 Chr.8: 1771465-1771465 on Build GRCh38
rs1008005551 Chr.8: 1771649-1771649 on Build GRCh38
rs972170426 Chr.8: 1771467-1771467 on Build GRCh38
rs1026460980 Chr.8: 1771477-1771477 on Build GRCh38
rs386834126 Chr.8: 1771374-1771374 on Build GRCh38
rs181971599 Chr.8: 1771444-1771444 on Build GRCh38
rs763671368 Chr.8: 1771267-1771267 on Build GRCh38
rs1010847109 Chr.8: 1771396-1771396 on Build GRCh38
rs374165512 Chr.8: 1771637-1771637 on Build GRCh38
rs998922998 Chr.8: 1771292-1771292 on Build GRCh38
rs780923228 Chr.8: 1771302-1771302 on Build GRCh38
rs764468452 Chr.8: 1771505-1771505 on Build GRCh38
rs771758079 Chr.8: 1771635-1771635 on Build GRCh38
rs1022265592 Chr.8: 1771403-1771403 on Build GRCh38
rs915726626 Chr.8: 1771654-1771654 on Build GRCh38
rs781115705 Chr.8: 1771512-1771512 on Build GRCh38
rs948587300 Chr.8: 1771656-1771656 on Build GRCh38
rs770109314 Chr.8: 1771420-1771420 on Build GRCh38
rs748077420 Chr.8: 1771547-1771547 on Build GRCh38
rs201327850 Chr.8: 1771241-1771241 on Build GRCh38
rs386834131 Chr.8: 1771561-1771561 on Build GRCh38
rs386834125 Chr.8: 1771281-1771281 on Build GRCh38
rs139041775 Chr.8: 1771430-1771430 on Build GRCh38
rs764468501 Chr.8: 1771565-1771565 on Build GRCh38
rs368365607 Chr.8: 1771261-1771261 on Build GRCh38
rs759598033 Chr.8: 1771480-1771483 on Build GRCh38
rs533351136 Chr.8: 1771342-1771342 on Build GRCh38
rs758707781 Chr.8: 1771501-1771501 on Build GRCh38
rs796052364 Chr.8: 1771230-1771230 on Build GRCh38
rs751170444 Chr.8: 1771283-1771283 on Build GRCh38
rs34030778 Chr.8: 1771328-1771328 on Build GRCh38
rs538101354 Chr.8: 1771608-1771608 on Build GRCh38
rs374031055 Chr.8: 1771557-1771557 on Build GRCh38
rs377273687 Chr.8: 1771387-1771387 on Build GRCh38
rs386834124 Chr.8: 1771263-1771263 on Build GRCh38
rs188259026 Chr.8: 1771563-1771563 on Build GRCh38
rs386834130 Chr.8: 1771527-1771527 on Build GRCh38
rs386834128 Chr.8: 1771518-1771518 on Build GRCh38
rs768139689 Chr.8: 1771577-1771577 on Build GRCh38
rs187297173 Chr.8: 1771460-1771460 on Build GRCh38
rs373957283 Chr.8: 1771254-1771254 on Build GRCh38
rs778862860 Chr.8: 1771621-1771621 on Build GRCh38
rs1044057107 Chr.8: 1771627-1771627 on Build GRCh38
rs982453047 Chr.8: 1771606-1771606 on Build GRCh38
rs749542144 Chr.8: 1771533-1771533 on Build GRCh38
rs750883349 Chr.8: 1771586-1771586 on Build GRCh38
rs755276576 Chr.8: 1771511-1771511 on Build GRCh38
rs781099399 Chr.8: 1771542-1771542 on Build GRCh38
rs571617007 Chr.8: 1771440-1771440 on Build GRCh38
rs386834123 Chr.8: 1771234-1771236 on Build GRCh38
rs776409703 Chr.8: 1771260-1771260 on Build GRCh38
rs143918546 Chr.8: 1771334-1771334 on Build GRCh38
rs878855052 Chr.8: 1771327-1771327 on Build GRCh38
rs966203442 Chr.8: 1771276-1771276 on Build GRCh38
rs907187780 Chr.8: 1771485-1771485 on Build GRCh38
rs535963507 Chr.8: 1771462-1771462 on Build GRCh38
rs753682420 Chr.8: 1771348-1771348 on Build GRCh38
rs753009900 Chr.8: 1771429-1771429 on Build GRCh38
rs143694317 Chr.8: 1771439-1771439 on Build GRCh38
rs748573529 Chr.8: 1771321-1771321 on Build GRCh38
rs879060656 Chr.8: 1771383-1771383 on Build GRCh38
rs774328095 Chr.8: 1771546-1771546 on Build GRCh38
rs769069643 Chr.8: 1771214-1771214 on Build GRCh38
rs1031756072 Chr.8: 1771513-1771513 on Build GRCh38
rs750162094 Chr.8: 1771349-1771349 on Build GRCh38
rs138581191 Chr.8: 1771509-1771509 on Build GRCh38
rs148124161 Chr.8: 1771378-1771378 on Build GRCh38
rs755248868 Chr.8: 1771310-1771310 on Build GRCh38
rs863224859 Chr.8: 1771266-1771266 on Build GRCh38
rs753439965 Chr.8: 1771487-1771487 on Build GRCh38
rs149308952 Chr.8: 1771524-1771524 on Build GRCh38
rs771203701 Chr.8: 1771258-1771258 on Build GRCh38
rs769440513 Chr.8: 1771339-1771339 on Build GRCh38
rs561448346 Chr.8: 1771324-1771324 on Build GRCh38
rs761348135 Chr.8: 1771352-1771352 on Build GRCh38
rs765097897 Chr.8: 1771262-1771262 on Build GRCh38
rs755831055 Chr.8: 1771357-1771357 on Build GRCh38
rs750004229 Chr.8: 1771356-1771356 on Build GRCh38
rs777819575 Chr.8: 1771330-1771330 on Build GRCh38
rs148417620 Chr.8: 1771576-1771576 on Build GRCh38
rs780551031 Chr.8: 1771213-1771213 on Build GRCh38
rs752026391 Chr.8: 1771506-1771506 on Build GRCh38
rs948684101 Chr.8: 1771453-1771453 on Build GRCh38
rs575820157 Chr.8: 1771255-1771255 on Build GRCh38
rs1011720410 Chr.8: 1771535-1771535 on Build GRCh38
rs568852954 Chr.8: 1771417-1771417 on Build GRCh38
rs1045641013 Chr.8: 1771667-1771667 on Build GRCh38
rs796052365 Chr.8: 1771359-1771359 on Build GRCh38
rs144495588 Chr.8: 1771553-1771553 on Build GRCh38
rs747882973 Chr.8: 1771631-1771631 on Build GRCh38
rs142269885 Chr.8: 1771428-1771428 on Build GRCh38

View Full Product Details