Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 1781392-1781890 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLN8
Entrez Gene ID
2055
Gene Name
ceroid-lipofuscinosis, neuronal 8
Gene Aliases
C8orf61, EPMR
UniGene
Hs.127675
Gene Chromosome Location
Chr.8: 1755778-1786570 on Build GRCh38
Transcripts
NM_018941.3

SNP Detail

SNP ID Chromosome Location
rs1002112658 Chr.8: 1781747-1781747 on Build GRCh38
rs371531678 Chr.8: 1781452-1781452 on Build GRCh38
rs925921787 Chr.8: 1781855-1781855 on Build GRCh38
rs141641935 Chr.8: 1781417-1781418 on Build GRCh38
rs936509740 Chr.8: 1781611-1781611 on Build GRCh38
rs1027031847 Chr.8: 1781435-1781435 on Build GRCh38
rs138680632 Chr.8: 1781499-1781499 on Build GRCh38
rs572140070 Chr.8: 1781774-1781774 on Build GRCh38
rs145067893 Chr.8: 1781681-1781681 on Build GRCh38
rs544980975 Chr.8: 1781646-1781646 on Build GRCh38
rs766771235 Chr.8: 1781578-1781578 on Build GRCh38
rs979079959 Chr.8: 1781849-1781849 on Build GRCh38
rs1013968509 Chr.8: 1781696-1781696 on Build GRCh38
rs1036502647 Chr.8: 1781518-1781518 on Build GRCh38
rs899300624 Chr.8: 1781712-1781712 on Build GRCh38
rs959850998 Chr.8: 1781453-1781453 on Build GRCh38
rs763555564 Chr.8: 1781447-1781447 on Build GRCh38
rs932037017 Chr.8: 1781496-1781496 on Build GRCh38
rs756284521 Chr.8: 1781864-1781864 on Build GRCh38
rs903749796 Chr.8: 1781420-1781421 on Build GRCh38
rs980255724 Chr.8: 1781439-1781439 on Build GRCh38
rs770805593 Chr.8: 1781607-1781607 on Build GRCh38
rs886506671 Chr.8: 1781655-1781655 on Build GRCh38
rs898003972 Chr.8: 1781546-1781546 on Build GRCh38
rs529235902 Chr.8: 1781426-1781426 on Build GRCh38
rs1034041248 Chr.8: 1781654-1781654 on Build GRCh38
rs963963716 Chr.8: 1781490-1781490 on Build GRCh38
rs573806537 Chr.8: 1781558-1781558 on Build GRCh38
rs900437098 Chr.8: 1781631-1781631 on Build GRCh38
rs917638190 Chr.8: 1781536-1781536 on Build GRCh38
rs556208557 Chr.8: 1781619-1781619 on Build GRCh38
rs548995371 Chr.8: 1781437-1781437 on Build GRCh38
rs553885210 Chr.8: 1781554-1781554 on Build GRCh38
rs752933812 Chr.8: 1781844-1781844 on Build GRCh38
rs534444139 Chr.8: 1781519-1781519 on Build GRCh38
rs1005008863 Chr.8: 1781668-1781668 on Build GRCh38
rs945922182 Chr.8: 1781465-1781465 on Build GRCh38
rs966423873 Chr.8: 1781821-1781821 on Build GRCh38
rs375844751 Chr.8: 1781820-1781820 on Build GRCh38
rs538172764 Chr.8: 1781462-1781462 on Build GRCh38
rs576653442 Chr.8: 1781475-1781475 on Build GRCh38
rs113649874 Chr.8: 1781609-1781609 on Build GRCh38
rs767805326 Chr.8: 1781822-1781822 on Build GRCh38
rs978706996 Chr.8: 1781466-1781466 on Build GRCh38
rs1024634397 Chr.8: 1781814-1781814 on Build GRCh38
rs960294203 Chr.8: 1781778-1781778 on Build GRCh38
rs933277288 Chr.8: 1781640-1781640 on Build GRCh38
rs397698661 Chr.8: 1781422-1781423 on Build GRCh38
rs576248925 Chr.8: 1781622-1781622 on Build GRCh38
rs150128397 Chr.8: 1781459-1781459 on Build GRCh38
rs1016458617 Chr.8: 1781485-1781485 on Build GRCh38
rs888100338 Chr.8: 1781579-1781579 on Build GRCh38
rs5888910 Chr.8: 1781416-1781417 on Build GRCh38
rs913083213 Chr.8: 1781463-1781463 on Build GRCh38
rs978326101 Chr.8: 1781564-1781564 on Build GRCh38
rs879732117 Chr.8: 1781695-1781695 on Build GRCh38
rs925945366 Chr.8: 1781487-1781487 on Build GRCh38
rs1020038331 Chr.8: 1781800-1781800 on Build GRCh38
rs890569373 Chr.8: 1781506-1781506 on Build GRCh38
rs1133517 Chr.8: 1781667-1781667 on Build GRCh38
rs953485767 Chr.8: 1781860-1781860 on Build GRCh38
rs980417085 Chr.8: 1781494-1781494 on Build GRCh38
rs571744333 Chr.8: 1781507-1781507 on Build GRCh38

View Full Product Details