Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 1784824-1785340 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLN8
Entrez Gene ID
2055
Gene Name
ceroid-lipofuscinosis, neuronal 8
Gene Aliases
C8orf61, EPMR
UniGene
Hs.127675
Gene Chromosome Location
Chr.8: 1755778-1786570 on Build GRCh38
Transcripts
NM_018941.3

SNP Detail

SNP ID Chromosome Location
rs538474232 Chr.8: 1785153-1785153 on Build GRCh38
rs898541992 Chr.8: 1784857-1784857 on Build GRCh38
rs577703425 Chr.8: 1784992-1784992 on Build GRCh38
rs969335570 Chr.8: 1785049-1785049 on Build GRCh38
rs575260928 Chr.8: 1784964-1784964 on Build GRCh38
rs1010895378 Chr.8: 1785079-1785079 on Build GRCh38
rs912377314 Chr.8: 1785117-1785117 on Build GRCh38
rs572839992 Chr.8: 1784843-1784843 on Build GRCh38
rs1010424880 Chr.8: 1785167-1785167 on Build GRCh38
rs577833196 Chr.8: 1785165-1785165 on Build GRCh38
rs139270315 Chr.8: 1784994-1784994 on Build GRCh38
rs184340330 Chr.8: 1784983-1784983 on Build GRCh38
rs149632384 Chr.8: 1785089-1785089 on Build GRCh38
rs1043265433 Chr.8: 1785177-1785177 on Build GRCh38
rs1002967998 Chr.8: 1785045-1785045 on Build GRCh38
rs919822538 Chr.8: 1785133-1785133 on Build GRCh38
rs56270086 Chr.8: 1785203-1785203 on Build GRCh38
rs530361554 Chr.8: 1784906-1784906 on Build GRCh38
rs982527937 Chr.8: 1785312-1785312 on Build GRCh38
rs1017858354 Chr.8: 1785216-1785216 on Build GRCh38
rs1047226814 Chr.8: 1785302-1785302 on Build GRCh38
rs563495055 Chr.8: 1784929-1784929 on Build GRCh38
rs538692429 Chr.8: 1785271-1785271 on Build GRCh38
rs972514446 Chr.8: 1785257-1785257 on Build GRCh38
rs555700725 Chr.8: 1784961-1784961 on Build GRCh38
rs1016807838 Chr.8: 1785037-1785037 on Build GRCh38
rs138156046 Chr.8: 1785033-1785033 on Build GRCh38
rs192236968 Chr.8: 1784897-1784897 on Build GRCh38
rs139385174 Chr.8: 1785141-1785141 on Build GRCh38
rs931245563 Chr.8: 1785137-1785137 on Build GRCh38
rs546551795 Chr.8: 1785287-1785287 on Build GRCh38
rs1042738175 Chr.8: 1784948-1784948 on Build GRCh38
rs550903191 Chr.8: 1784905-1784905 on Build GRCh38
rs143200025 Chr.8: 1785267-1785267 on Build GRCh38
rs375136015 Chr.8: 1785100-1785100 on Build GRCh38
rs1019383939 Chr.8: 1785229-1785229 on Build GRCh38
rs992064606 Chr.8: 1784892-1784892 on Build GRCh38
rs913292115 Chr.8: 1785310-1785310 on Build GRCh38
rs542766598 Chr.8: 1785060-1785060 on Build GRCh38
rs111861349 Chr.8: 1785316-1785316 on Build GRCh38
rs995542698 Chr.8: 1784860-1784860 on Build GRCh38
rs140524201 Chr.8: 1785010-1785010 on Build GRCh38
rs961439738 Chr.8: 1785248-1785248 on Build GRCh38
rs75237112 Chr.8: 1785179-1785179 on Build GRCh38
rs931515777 Chr.8: 1785230-1785230 on Build GRCh38
rs761105938 Chr.8: 1784861-1784861 on Build GRCh38
rs549509252 Chr.8: 1785138-1785138 on Build GRCh38
rs55915521 Chr.8: 1785091-1785091 on Build GRCh38
rs533136126 Chr.8: 1785103-1785103 on Build GRCh38
rs865843802 Chr.8: 1785264-1785264 on Build GRCh38
rs914121104 Chr.8: 1784932-1784932 on Build GRCh38
rs910827391 Chr.8: 1785122-1785122 on Build GRCh38
rs946135394 Chr.8: 1785313-1785313 on Build GRCh38
rs566854151 Chr.8: 1784946-1784946 on Build GRCh38
rs1057457670 Chr.8: 1785297-1785297 on Build GRCh38
rs542056790 Chr.8: 1784925-1784925 on Build GRCh38
rs566792763 Chr.8: 1785126-1785126 on Build GRCh38
rs146897459 Chr.8: 1785099-1785099 on Build GRCh38
rs533268381 Chr.8: 1785284-1785284 on Build GRCh38
rs931339390 Chr.8: 1785008-1785008 on Build GRCh38
rs557553625 Chr.8: 1785154-1785154 on Build GRCh38
rs573934874 Chr.8: 1785198-1785198 on Build GRCh38
rs1035065538 Chr.8: 1785187-1785187 on Build GRCh38
rs553233764 Chr.8: 1784935-1784935 on Build GRCh38
rs956455736 Chr.8: 1784890-1784890 on Build GRCh38
rs188634468 Chr.8: 1785102-1785102 on Build GRCh38
rs546907678 Chr.8: 1785114-1785114 on Build GRCh38
rs558540267 Chr.8: 1785294-1785294 on Build GRCh38
rs906025374 Chr.8: 1784875-1784875 on Build GRCh38
rs964984529 Chr.8: 1785265-1785265 on Build GRCh38
rs868678220 Chr.8: 1785286-1785286 on Build GRCh38
rs118000123 Chr.8: 1784889-1784889 on Build GRCh38
rs947029048 Chr.8: 1784945-1784945 on Build GRCh38
rs1036226676 Chr.8: 1785063-1785063 on Build GRCh38
rs1039450327 Chr.8: 1784967-1784967 on Build GRCh38
rs184354344 Chr.8: 1785196-1785196 on Build GRCh38
rs941214555 Chr.8: 1785157-1785157 on Build GRCh38
rs150391413 Chr.8: 1785018-1785018 on Build GRCh38
rs11995303 Chr.8: 1785253-1785253 on Build GRCh38
rs190471975 Chr.8: 1785268-1785268 on Build GRCh38
rs537547112 Chr.8: 1784979-1784979 on Build GRCh38
rs979611112 Chr.8: 1785204-1785204 on Build GRCh38
rs564215205 Chr.8: 1785272-1785272 on Build GRCh38
rs117588996 Chr.8: 1784958-1784958 on Build GRCh38
rs996438793 Chr.8: 1785193-1785193 on Build GRCh38
rs192186917 Chr.8: 1785127-1785127 on Build GRCh38
rs116856439 Chr.8: 1785034-1785034 on Build GRCh38
rs541741154 Chr.8: 1784883-1784883 on Build GRCh38
rs919682468 Chr.8: 1785217-1785217 on Build GRCh38
rs932407980 Chr.8: 1784936-1784936 on Build GRCh38
rs939931356 Chr.8: 1784970-1784970 on Build GRCh38
rs1003432367 Chr.8: 1784882-1784882 on Build GRCh38
rs892001225 Chr.8: 1785069-1785069 on Build GRCh38
rs905876715 Chr.8: 1785140-1785140 on Build GRCh38

View Full Product Details