The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|---|
RefSeq | NM_001085354.2 | NP_001078823.1 | 5 - 6 | 945 | 133 | |
NM_024657.4 | NP_078933.3 | 5 - 6 | 945 | 133 | ||
XM_005262190.3 | XP_005262247.1 | 4 - 5 | 835 | 133 | ||
XM_011531027.2 | XP_011529329.1 | 2 - 3 | 233 | 133 | ||
XM_017029844.1 | XP_016885333.1 | 4 - 5 | 835 | 133 | ||
GenBank mRNA | AK056235.1 | - | 1 - 2 | 157 | 133 | |
AK300305.1 | - | 4 - 5 | 520 | 133 | ||
AY927537.1 | - | LIT3543 AY927537 |
3 - 4 | 480 | 133 | |
EF125209.1 | - | 5 - 6 | 670 | 133 |