Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MORC4
Entrez Gene ID
79710
Gene Name
MORC family CW-type zinc finger 4
Gene Aliases
ZCW4, ZCWCC2, dJ75H8.2
Chromosome Location
Chr.X: 106940300 - 107000244 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085354.2 NP_001078823.1 15 - 16 2658 70
NM_024657.4 NP_078933.3 15 - 16 2658 70
XM_005262190.3 XP_005262247.1 14 - 15 2548 70
XM_006724691.2 XP_006724754.1 13 - 14 2292 70
XM_011531027.2 XP_011529329.1 12 - 13 1946 70
XM_017029844.1 XP_016885333.1 14 - 15 2548 70
GenBank mRNA AK021627.1 - 4 - 5 972 70
AK056235.1 - 11 - 12 1870 70
AK300305.1 - 14 - 15 2233 70
EF125209.1 - 15 - 16 2383 70
EF125210.1 - 14 - 15 2442 70

View Full Product Details