Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLFN5
Entrez Gene ID
162394
Gene Name
schlafen family member 5
Gene Aliases
-
Chromosome Location
Chr.17: 35243026 - 35273650 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144975.3 NP_659412.3 1 - 2 77 56
XM_005257934.3 XP_005257991.1 1 - 2 118 56
GenBank mRNA AK054668.1 - 1 - 2 72 56
AK303299.1 - 1 - 2 72 56
AK303632.1 - 1 - 2 74 56
BC021238.1 - 1 - 2 55 56
BP281007.1 - 1 - 2 77 56
BX647942.1 - 1 - 2 74 56
CD704489.1 - 1 - 2 87 56

View Full Product Details