The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256054.2 | NP_001242983.1 | 2 - 3 | 641 | 123 |
NM_018325.4 | NP_060795.1 | 2 - 3 | 546 | 123 | |
NM_145005.6 | NP_659442.2 | 2 - 3 | 563 | 123 | |
GenBank mRNA | AK057806.1 | - | 2 - 3 | 541 | 123 |
AK291425.1 | - | 2 - 3 | 536 | 123 | |
AL832229.1 | - | 1 - 2 | 503 | 123 | |
AL832467.1 | - | 2 - 3 | 502 | 123 | |
BC020851.2 | - | 1 - 2 | 483 | 123 | |
BC068445.1 | - | 1 - 2 | 495 | 123 | |
BG718065.1 | - | 2 - 3 | 552 | 123 | |
BG722125.1 | - | 2 - 3 | 540 | 124 | |
BQ068108.1 | - | - | 652 | 124 | |
JN681271.1 | - | 2 - 3 | 619 | 123 |