Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OSCP1
Entrez Gene ID
127700
Gene Name
organic solute carrier partner 1
Gene Aliases
C1orf102, NOR1
Chromosome Location
Chr.1: 36417906 - 36450485 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145047.4 NP_659484.4 1 - 2 228 115
NM_206837.2 NP_996668.1 1 - 2 228 115
XM_017000303.1 XP_016855792.1 1 - 2 164 115
XM_017000304.1 XP_016855793.1 1 - 2 164 115
GenBank mRNA AB079075.1 - 1 - 2 112 115
AF462348.1 - 1 - 2 270 115
BC018069.1 - 1 - 2 194 115
BU854079.1 - - 189 115

View Full Product Details