The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145648.3 | NP_663623.1 | 5 - 6 | 1312 | 103 |
XM_011537895.1 | XP_011536197.1 | 5 - 6 | 1432 | 103 | |
XM_017018791.1 | XP_016874280.1 | 5 - 6 | 1432 | 103 | |
XM_017018792.1 | XP_016874281.1 | 5 - 6 | 1432 | 103 | |
XM_017018793.1 | XP_016874282.1 | 5 - 6 | 1282 | 103 | |
GenBank mRNA | AF461893.1 | - | 3 - 4 | 434 | 103 |
AK095717.1 | - | 4 - 5 | 926 | 103 | |
AY038999.1 | - | 5 - 6 | 1317 | 103 | |
AY050629.1 | - | 5 - 6 | 1262 | 103 | |
BC028394.2 | - | 5 - 6 | 1065 | 103 | |
BC146803.1 | - | 5 - 6 | 1288 | 103 |