The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_147196.2 | NP_671729.2 | 2 - 3 | 363 | 75 |
XM_006713097.3 | XP_006713160.1 | 2 - 3 | 934 | 75 | |
GenBank mRNA | AY081842.1 | - | 2 - 3 | 363 | 75 |
BC126258.1 | - | 2 - 3 | 326 | 75 | |
BC126260.1 | - | 2 - 3 | 326 | 75 |