The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005300.3 | NP_005291.1 | 1 - 2 | 115 | 138 |
XM_005272597.3 | XP_005272654.1 | 1 - 2 | 2545 | 138 | |
GenBank mRNA | BC020678.1 | - | 1 - 2 | 82 | 138 |