The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042481.2 | NP_001035946.1 | 13 - 14 | 1973 | 67 |
NM_001267046.1 | NP_001253975.1 | 12 - 13 | 1773 | 67 | |
NM_001267047.1 | NP_001253976.1 | 3 - 4 | 530 | 67 | |
NM_152330.3 | NP_689543.1 | 12 - 13 | 1749 | 67 | |
XM_006720029.1 | XP_006720092.1 | 12 - 13 | 1870 | 67 | |
XM_006720030.1 | XP_006720093.1 | 12 - 13 | 1846 | 67 | |
XM_011536423.1 | XP_011534725.1 | 13 - 14 | 2017 | 67 | |
XM_011536424.1 | XP_011534726.1 | 14 - 15 | 2142 | 67 | |
XM_017020967.1 | XP_016876456.1 | 12 - 13 | 1723 | 67 | |
XM_017020968.1 | XP_016876457.1 | 12 - 13 | 1699 | 67 | |
GenBank mRNA | AK055545.1 | - | 12 - 13 | 1684 | 67 |
AK126521.1 | - | 10 - 11 | 1394 | 67 | |
AK315764.1 | - | 12 - 13 | 1670 | 67 | |
AL833158.1 | - | 13 - 14 | 1959 | 67 | |
AL833380.1 | - | 7 - 8 | 988 | 67 | |
AL833661.1 | - | 7 - 8 | 969 | 67 | |
BC020521.2 | - | 12 - 13 | 1697 | 67 | |
BC029870.1 | - | 3 - 4 | 530 | 67 | |
BX161430.1 | - | 12 - 13 | 1749 | 67 | |
BX647239.1 | - | 3 - 4 | 2318 | 67 | |
BX647863.1 | - | 12 - 13 | 1661 | 67 | |
BX648295.1 | - | 3 - 4 | 2319 | 67 |