The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003876.2 | NP_003867.1 | 1 - 2 | 509 | 73 |
GenBank mRNA | AK311965.1 | - | 1 - 2 | 75 | 73 |
BC002819.2 | - | 1 - 2 | 97 | 73 | |
BC005268.1 | - | 1 - 2 | 509 | 73 | |
BC020815.1 | - | 1 - 2 | 75 | 73 | |
BT006768.1 | - | 1 - 2 | 66 | 73 | |
X51804.1 | - | 1 - 2 | 324 | 73 |