Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IQCF3
Entrez Gene ID
401067
Gene Name
IQ motif containing F3
Gene Aliases
-
Chromosome Location
Chr.3: 51826883 - 51830858 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC021188.2 - 2 - 3 101 70

View Full Product Details