The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145166.3 | NP_660149.2 | 4 - 5 | 2018 | 73 |
GenBank mRNA | AK127065.1 | - | 5 - 6 | 1712 | 73 |
AL133062.1 | - | 3 - 4 | 711 | 73 | |
BC021855.1 | - | 3 - 4 | 676 | 73 |