The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182935.3 | NP_891980.1 | 3 - 4 | 462 | 95 |
NR_003090.2 | 3 - 4 | 462 | 95 | ||
GenBank mRNA | BC022471.1 | - | 3 - 4 | 356 | 95 |
BC075796.1 | - | 3 - 4 | 430 | 95 | |
BC119694.1 | - | 2 - 3 | 239 | 95 | |
BC119695.1 | - | 2 - 3 | 239 | 95 | |
DA343382.1 | - | 3 - 4 | 356 | 95 | |
DA418624.1 | - | 3 - 4 | 456 | 95 | |
EB387411.1 | - | 3 - 4 | 330 | 95 |