The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001131065.1 | NP_001124537.1 | 3 - 4 | 1491 | 102 |
NM_001131066.1 | NP_001124538.1 | 3 - 4 | 336 | 102 | |
XM_011543360.2 | XP_011541662.1 | 4 - 5 | 1379 | 102 | |
XM_017009413.1 | XP_016864902.1 | 3 - 4 | 503 | 102 | |
GenBank mRNA | BC024023.1 | - | 1 - 2 | 110 | 102 |
BI551847.1 | - | 3 - 4 | 339 | 102 | |
BM263054.1 | - | 2 - 3 | 300 | 102 |