The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015082.1 | NP_055897.1 | 5 - 6 | 819 | 81 |
XM_011543283.1 | XP_011541585.1 | 5 - 6 | 726 | 81 | |
XM_011543284.1 | XP_011541586.1 | 5 - 6 | 797 | 81 | |
XM_011543286.2 | XP_011541588.1 | 2 - 3 | 280 | 81 | |
GenBank mRNA | AB028984.1 | - | 1 - 2 | 156 | 81 |
AK055684.1 | - | 5 - 6 | 797 | 81 | |
BC024300.1 | - | 5 - 6 | 851 | 81 |