The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015082.1 | NP_055897.1 | 12 - 13 | 1678 | 78 |
XM_011543283.1 | XP_011541585.1 | 12 - 13 | 1585 | 78 | |
XM_011543284.1 | XP_011541586.1 | 11 - 12 | 1629 | 78 | |
XM_011543286.2 | XP_011541588.1 | 9 - 10 | 1139 | 78 | |
XM_011543287.2 | XP_011541589.1 | 6 - 7 | 1367 | 78 | |
XM_017009251.1 | XP_016864740.1 | 7 - 8 | 1394 | 78 | |
GenBank mRNA | AB028984.1 | - | 8 - 9 | 1015 | 78 |
AK056284.1 | - | 4 - 5 | 678 | 78 | |
BC024300.1 | - | 12 - 13 | 1710 | 78 | |
BX640845.1 | - | 10 - 11 | 1093 | 78 |