Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTC8
Entrez Gene ID
123016
Gene Name
tetratricopeptide repeat domain 8
Gene Aliases
BBS8, RP51
Chromosome Location
Chr.14: 88824153 - 88877996 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001288781.1 NP_001275710.1 9 - 10 949 87
NM_001288782.1 NP_001275711.1 7 - 8 869 87
NM_001288783.1 NP_001275712.1 8 - 9 841 87
NM_144596.3 NP_653197.2 9 - 10 901 87
NM_198309.3 NP_938051.1 8 - 9 871 87
NM_198310.3 NP_938052.1 7 - 8 781 87
XM_006720035.1 XP_006720098.1 7 - 8 1153 87
XM_006720037.2 XP_006720100.1 6 - 7 1063 87
XM_011536432.1 XP_011534734.1 8 - 9 1231 87
XM_011536433.1 XP_011534735.1 8 - 9 1231 87
XM_011536434.1 XP_011534736.1 7 - 8 1141 87
XM_011536435.2 XP_011534737.1 7 - 8 617 87
XM_017020977.1 XP_016876466.1 7 - 8 1153 87
XM_017020978.1 XP_016876467.1 6 - 7 1063 87
GenBank mRNA AK093891.1 - 7 - 8 719 87
AK124675.1 - 8 - 9 751 87
AL833901.1 - 6 - 7 605 87
AY366523.1 - 7 - 8 727 87
AY366524.1 - 6 - 7 637 87
AY373972.1 - 7 - 8 727 87
BC001563.1 - 3 - 4 219 87
BC026351.1 - 8 - 9 805 87
BC095433.1 - 7 - 8 709 87
BX161472.1 - 6 - 7 709 87
BX248248.1 - 4 - 5 313 87
KU178849.1 - 8 - 9 753 87
KU178850.1 - 7 - 8 675 87
KU178851.1 - 6 - 7 585 87
KU178852.1 - 2 - 3 195 87

View Full Product Details