Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C9orf43
Entrez Gene ID
257169
Gene Name
chromosome 9 open reading frame 43
Gene Aliases
-
Chromosome Location
Chr.9: 113410644 - 113429684 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152786.2 NP_689999.1 1 - 2 433 92
GenBank mRNA BC026884.1 - 1 - 2 399 92
DB048582.1 - 1 - 2 433 92

View Full Product Details