Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C9orf43
Entrez Gene ID
257169
Gene Name
chromosome 9 open reading frame 43
Gene Aliases
-
Chromosome Location
Chr.9: 113410644 - 113429684 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278629.1 NP_001265558.1 13 - 14 1581 78
NM_001278630.1 NP_001265559.1 13 - 14 1438 78
NM_152786.2 NP_689999.1 13 - 14 1654 78
XM_005251885.3 XP_005251942.1 11 - 12 960 78
XM_011518479.2 XP_011516781.1 11 - 12 1236 78
GenBank mRNA AK301872.1 - 13 - 14 1441 78
AK313867.1 - 13 - 14 1570 78
BC026884.1 - 13 - 14 1620 78
BC033037.2 - 13 - 14 1539 78

View Full Product Details