The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015056.2 | NP_055871.1 | 5 - 6 | 530 | 71 |
GenBank mRNA | AK056263.1 | - | 5 - 6 | 493 | 71 |
BC014005.2 | - | 5 - 6 | 502 | 71 | |
BC028386.2 | - | 5 - 6 | 535 | 71 | |
D80001.1 | - | 5 - 6 | 428 | 71 |