The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013848.3 | NP_001013870.1 | 15 - 16 | 1540 | 128 |
NM_001319194.1 | NP_001306123.1 | 15 - 16 | 1606 | 128 | |
NM_001319200.1 | NP_001306129.1 | 10 - 11 | 1185 | 128 | |
NM_019053.5 | NP_061926.3 | 14 - 15 | 1494 | 128 | |
XM_017016345.1 | XP_016871834.1 | 14 - 15 | 1575 | 128 | |
XM_017016347.1 | XP_016871836.1 | 14 - 15 | 1778 | 128 | |
XM_017016348.1 | XP_016871837.1 | 15 - 16 | 1601 | 128 | |
GenBank mRNA | AK002113.1 | - | 6 - 7 | 492 | 128 |
AK123268.1 | - | 1 - 2 | 849 | 128 | |
AK128190.1 | - | 15 - 16 | 1540 | 128 | |
AK293857.1 | - | 14 - 15 | 1457 | 128 | |
AK300285.1 | - | 15 - 16 | 1606 | 128 | |
AK315544.1 | - | 14 - 15 | 1429 | 128 | |
AL137438.1 | - | 9 - 10 | 791 | 128 | |
BC028395.2 | - | 14 - 15 | 1429 | 128 | |
EF571007.1 | - | 10 - 11 | 1126 | 128 |