Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VSTM2A
Entrez Gene ID
222008
Gene Name
V-set and transmembrane domain containing 2A
Gene Aliases
VSTM2
Chromosome Location
Chr.7: 54542064 - 54571080 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301009.1 NP_001287938.1 3 - 4 701 126
NM_001317843.1 NP_001304772.1 3 - 4 701 126
NM_182546.3 NP_872352.2 3 - 4 701 126
XM_006715665.3 XP_006715728.1 3 - 4 962 126
XM_006715666.3 XP_006715729.1 3 - 4 962 126
XM_017011841.1 XP_016867330.1 3 - 4 962 126
GenBank mRNA BC028404.1 - 3 - 4 576 126
BI553644.1 - 3 - 4 508 126
BI754879.1 - 3 - 4 241 126
BI758069.1 - 3 - 4 363 126
DN991717.1 - 3 - 4 463 126

View Full Product Details