The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080414.3 | NP_001073883.2 | 29 - 30 | 5211 | 78 |
XM_011536796.2 | XP_011535098.1 | 29 - 30 | 5165 | 78 | |
XM_017021336.1 | XP_016876825.1 | 14 - 15 | 2274 | 78 | |
GenBank mRNA | AK022593.1 | - | 2 - 3 | 400 | 78 |
BC028565.2 | - | 1 - 2 | 86 | 78 | |
BC035914.1 | - | 5 - 6 | 923 | 78 | |
BX248302.1 | - | 5 - 6 | 869 | 78 |