Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TDRD7
Entrez Gene ID
23424
Gene Name
tudor domain containing 7
Gene Aliases
CATC4, PCTAIRE2BP, TRAP
Chromosome Location
Chr.9: 97412020 - 97496125 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001302884.1 NP_001289813.1 3 - 4 574 76
NM_014290.2 NP_055105.2 4 - 5 787 76
XM_006717026.3 XP_006717089.1 4 - 5 664 76
GenBank mRNA AK294488.1 - 3 - 4 500 76
AK314853.1 - 4 - 5 699 76
BC028694.1 - 4 - 5 786 76

View Full Product Details