The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018082.2 | NP_001018092.1 | 2 - 3 | 404 | 72 |
GenBank mRNA | BC029594.1 | - | 2 - 3 | 404 | 72 |
BC119704.2 | - | 2 - 3 | 398 | 72 | |
BC119705.2 | - | 2 - 3 | 398 | 72 | |
BX105371.1 | - | 2 - 3 | 244 | 72 |