Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- MCTP1
- Entrez Gene ID
-
79772
- Gene Name
- multiple C2 and transmembrane domain containing 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.5: 94703690 - 95284575 on Build GRCh38
View Full Product Details