Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LPIN1
Entrez Gene ID
23175
Gene Name
lipin 1
Gene Aliases
PAP1
Chromosome Location
Chr.2: 11677544 - 11827409 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261427.1 NP_001248356.1 12 - 13 1905 87
NM_001261428.1 NP_001248357.1 13 - 14 1934 87
NM_145693.2 NP_663731.1 11 - 12 1695 87
XM_006711869.2 XP_006711932.1 12 - 13 1861 87
XM_006711870.3 XP_006711933.1 13 - 14 2100 87
XM_006711871.2 XP_006711934.1 12 - 13 1803 87
XM_006711872.2 XP_006711935.1 12 - 13 2027 87
XM_006711874.2 XP_006711937.1 13 - 14 3631 87
XM_011510333.2 XP_011508635.1 13 - 14 1966 87
XM_011510334.2 XP_011508636.1 13 - 14 1996 87
XM_011510335.2 XP_011508637.1 15 - 16 2459 87
XM_011510336.2 XP_011508638.1 14 - 15 2341 87
XM_017003623.1 XP_016859112.1 13 - 14 1833 87
XM_017003624.1 XP_016859113.1 13 - 14 2028 87
XM_017003625.1 XP_016859114.1 12 - 13 1745 87
XM_017003626.1 XP_016859115.1 12 - 13 1800 87
XM_017003627.1 XP_016859116.1 15 - 16 2456 87
XM_017003628.1 XP_016859117.1 14 - 15 2351 87
XM_017003629.1 XP_016859118.1 12 - 13 1920 87
XM_017003630.1 XP_016859119.1 11 - 12 1637 87
GenBank mRNA AK290235.1 - 11 - 12 1655 87
AK294742.1 - 13 - 14 1918 87
AK294853.1 - 8 - 9 1087 87
BC030537.1 - 11 - 12 1688 87
CR749806.1 - 7 - 8 1203 87
D80010.1 - 11 - 12 1638 87

View Full Product Details