Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MIS12
Entrez Gene ID
79003
Gene Name
MIS12, kinetochore complex component
Gene Aliases
2510025F08Rik, KNTC2AP, MTW1, hMis12
Chromosome Location
Chr.17: 5486374 - 5490814 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258218.1 NP_001245147.1 1 - 2 308 94
GenBank mRNA AK056072.1 - 1 - 2 286 94
BC000229.1 - 1 - 2 94 94

View Full Product Details