The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040437.2 | NP_001035527.1 | 4 - 5 | 617 | 60 |
NM_001040438.2 | NP_001035528.1 | 3 - 4 | 523 | 60 | |
NM_001287482.1 | NP_001274411.1 | 4 - 5 | 628 | 60 | |
NM_001287483.1 | NP_001274412.1 | 2 - 3 | 482 | 60 | |
NM_001287484.1 | NP_001274413.1 | 3 - 4 | 527 | 60 | |
NM_001287485.1 | NP_001274414.1 | 4 - 5 | 474 | 60 | |
NM_001287486.1 | NP_001274415.1 | 4 - 5 | 471 | 60 | |
NM_001287487.1 | NP_001274416.1 | 3 - 4 | 380 | 60 | |
NM_001287488.1 | NP_001274417.1 | 3 - 4 | 268 | 60 | |
GenBank mRNA | AA720680.1 | - | 1 - 2 | 60 | |
AJ249731.1 | - | 3 - 4 | 532 | 60 | |
AJ249732.1 | - | 2 - 3 | 491 | 60 | |
AL521625.3 | - | 4 - 5 | 603 | 60 | |
BC000706.1 | - | 3 - 4 | 504 | 60 | |
BC017660.1 | - | 4 - 5 | 421 | 60 | |
BC066330.1 | - | 3 - 4 | 522 | 60 | |
BG109213.1 | - | 4 - 5 | 545 | 60 | |
BJ996767.1 | - | 2 - 3 | 482 | 60 | |
BM690817.1 | - | 3 - 4 | 342 | 60 | |
BQ920527.1 | - | 4 - 5 | 601 | 60 | |
CD247791.1 | - | 4 - 5 | 490 | 60 | |
CD249104.1 | - | 3 - 4 | 538 | 60 | |
HY060373.1 | - | 3 - 4 | 266 | 60 | |
N91812.1 | - | 1 - 2 | 60 |