The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020531.2 | NP_065392.1 | 2 - 3 | 311 | 82 |
XM_005260763.3 | XP_005260820.1 | 2 - 3 | 452 | 82 | |
GenBank mRNA | AB033767.1 | - | 2 - 3 | 250 | 82 |
AK291129.1 | - | 2 - 3 | 82 | ||
AK301959.1 | - | 2 - 3 | 452 | 82 | |
AY359076.1 | - | 1 - 2 | 98 | 82 | |
BC003501.1 | - | 2 - 3 | 235 | 82 | |
BG773501.1 | - | 2 - 3 | 508 | 82 | |
CF131759.1 | - | 2 - 3 | 280 | 82 |