The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020209.3 | NP_064594.3 | 4 - 5 | 2181 | 103 |
GenBank mRNA | AK055673.1 | - | 4 - 5 | 1120 | 103 |
AK056268.1 | - | 4 - 5 | 2182 | 103 | |
AL390078.1 | - | 5 - 6 | 439 | 103 | |
BC007206.1 | - | 4 - 5 | 943 | 103 |