The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001323614.1 | NP_001310543.1 | 14 - 15 | 2321 | 60 |
NM_001323615.1 | NP_001310544.1 | 13 - 14 | 2183 | 60 | |
NM_001323616.1 | NP_001310545.1 | 15 - 16 | 2453 | 60 | |
NM_001323617.1 | NP_001310546.1 | 15 - 16 | 2288 | 60 | |
NM_015327.2 | NP_056142.2 | 14 - 15 | 2260 | 60 | |
XM_017000842.1 | XP_016856331.1 | 10 - 11 | 2206 | 60 | |
XM_017000843.1 | XP_016856332.1 | 13 - 14 | 2188 | 60 | |
GenBank mRNA | AB029012.2 | - | 14 - 15 | 2262 | 60 |
AB085691.1 | - | 14 - 15 | 2188 | 60 | |
AK027486.1 | - | 1 - 2 | 48 | 60 | |
AL137738.1 | - | 3 - 4 | 362 | 60 | |
AY168922.1 | - | 14 - 15 | 2114 | 60 | |
AY336728.1 | - | 14 - 15 | 2111 | 60 | |
BC007453.2 | - | 3 - 4 | 707 | 60 | |
BC038296.1 | - | 14 - 15 | 2261 | 60 |