The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001323614.1 | NP_001310543.1 | 16 - 17 | 2557 | 94 |
NM_001323615.1 | NP_001310544.1 | 16 - 17 | 2578 | 94 | |
NM_001323616.1 | NP_001310545.1 | 18 - 19 | 2848 | 94 | |
NM_001323617.1 | NP_001310546.1 | 18 - 19 | 2683 | 94 | |
NM_015327.2 | NP_056142.2 | 17 - 18 | 2655 | 94 | |
XM_017000842.1 | XP_016856331.1 | 13 - 14 | 2601 | 94 | |
XM_017000843.1 | XP_016856332.1 | 16 - 17 | 2583 | 94 | |
GenBank mRNA | AB029012.2 | - | 17 - 18 | 2657 | 94 |
AB085691.1 | - | 17 - 18 | 2583 | 94 | |
AK027486.1 | - | 4 - 5 | 443 | 94 | |
AL137738.1 | - | 6 - 7 | 757 | 94 | |
AY168922.1 | - | 17 - 18 | 2509 | 94 | |
AY336728.1 | - | 17 - 18 | 2506 | 94 | |
BC007453.2 | - | 6 - 7 | 1102 | 94 | |
BC038296.1 | - | 17 - 18 | 2656 | 94 |