Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NAAA
Entrez Gene ID
27163
Gene Name
N-acylethanolamine acid amidase
Gene Aliases
ASAHL, PLT
Chromosome Location
Chr.4: 75910655 - 75941013 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042402.1 NP_001035861.1 7 - 8 965 62
NM_014435.3 NP_055250.2 7 - 8 965 62
XM_005262920.3 XP_005262977.1 7 - 8 963 62
XM_005262921.3 XP_005262978.1 7 - 8 963 62
XM_005262923.2 XP_005262980.1 7 - 8 963 62
XM_005262924.2 XP_005262981.1 7 - 8 963 62
XM_005262925.3 XP_005262982.1 7 - 8 963 62
XM_006714180.2 XP_006714243.1 7 - 8 963 62
XM_011531852.2 XP_011530154.1 7 - 8 963 62
XM_011531854.2 XP_011530156.1 7 - 8 963 62
XM_017008027.1 XP_016863516.1 7 - 8 963 62
XM_017008028.1 XP_016863517.1 7 - 8 963 62
XM_017008029.1 XP_016863518.1 7 - 8 963 62
XM_017008030.1 XP_016863519.1 7 - 8 963 62
GenBank mRNA AB161353.1 - 7 - 8 901 62
AK024677.1 - - 1465 62
AK301132.1 - 6 - 7 790 62
BC006388.2 - 6 - 7 868 62
M92449.1 - 6 - 7 661 62
W47597.1 - 2 - 3 222 62

View Full Product Details