The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015138.4 | NP_055953.3 | 14 - 15 | 1754 | 84 |
GenBank mRNA | AK292302.1 | - | 14 - 15 | 1726 | 84 |
BC015052.2 | - | 14 - 15 | 1704 | 84 | |
D87440.2 | - | 14 - 15 | 1720 | 84 |