The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001284401.1 | NP_001271330.1 | 1 - 2 | 417 | 96 |
NM_138807.3 | NP_620162.1 | 1 - 2 | 417 | 96 | |
NR_104314.1 | 1 - 2 | 417 | 96 | ||
NR_135611.1 | 1 - 2 | 417 | 96 | ||
XM_005264873.3 | XP_005264930.1 | 1 - 2 | 417 | 96 | |
XM_005264875.3 | XP_005264932.1 | 1 - 2 | 417 | 96 | |
XM_017005725.1 | XP_016861214.1 | 1 - 2 | 417 | 96 | |
XM_017005726.1 | XP_016861215.1 | 1 - 2 | 417 | 96 | |
XM_017005727.1 | XP_016861216.1 | 1 - 2 | 398 | 96 | |
XM_017005728.1 | XP_016861217.1 | 1 - 2 | 398 | 96 | |
GenBank mRNA | AK092776.1 | - | 1 - 2 | 379 | 96 |
AK295839.1 | - | 1 - 2 | 277 | 96 | |
AK303598.1 | - | 1 - 2 | 390 | 96 | |
AK310993.1 | - | 1 - 2 | 343 | 96 | |
BC015088.2 | - | 1 - 2 | 376 | 96 | |
DB475191.1 | - | 1 - 2 | 419 | 96 |