The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114093.1 | NP_001107565.1 | 4 - 5 | 737 | 106 |
NM_015578.2 | NP_056393.2 | 4 - 5 | 737 | 106 | |
XM_005258719.4 | XP_005258776.1 | 4 - 5 | 756 | 106 | |
XM_005258721.4 | XP_005258778.1 | 4 - 5 | 756 | 106 | |
XM_017026577.1 | XP_016882066.1 | 4 - 5 | 756 | 106 | |
GenBank mRNA | AB069974.1 | - | 4 - 5 | 616 | 106 |
AB069975.1 | - | 4 - 5 | 616 | 106 | |
AK027369.1 | - | 3 - 4 | 347 | 106 | |
AK027643.1 | - | 4 - 5 | 618 | 106 | |
AK127401.1 | - | 1 - 2 | 860 | 106 | |
AL833221.1 | - | 2 - 3 | 210 | 106 | |
AL834398.1 | - | 4 - 5 | 646 | 106 | |
BC016842.1 | - | 4 - 5 | 737 | 106 |