The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300819.2 | NP_001287748.1 | 1 - 2 | 101 | 93 |
NM_001308130.1 | NP_001295059.1 | 1 - 2 | 101 | 93 | |
NM_017665.3 | NP_060135.1 | 1 - 2 | 101 | 93 | |
GenBank mRNA | AK000101.1 | - | 1 - 2 | 45 | 93 |
AK300543.1 | - | 1 - 2 | 69 | 93 | |
BC005211.1 | - | 1 - 2 | 58 | 93 | |
BI826979.1 | - | 1 - 2 | 101 | 93 | |
DA528172.1 | - | 1 - 2 | 45 | 93 |